Assay Details
| Gene Symbol: LOC100759640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI882615.1 | - | 3 - 4 | 311 | 88 | |
| JP047938.1 | - | 3 - 4 | 311 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759640| Gene Symbol: LOC100759640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI882615.1 | - | 3 - 4 | 311 | 88 | |
| JP047938.1 | - | 3 - 4 | 311 | 88 | ||