Assay Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007619130.1 | XP_007617320.1 | 7 - 8 | 987 | 69 | |
| GenBank mRNA | JI871264.1 | - | 7 - 8 | 949 | 69 | |
| JP041587.1 | - | 7 - 8 | 949 | 69 | ||
More Information
Set Membership: |
|

Rad21
Genomic Map