Assay Details
| Gene Symbol: Gk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648509.1 | XP_007646699.1 | 13 - 14 | 1075 | 68 | |
| XM_007648510.1 | XP_007646700.1 | 12 - 13 | 1057 | 68 | ||
| XM_007648511.1 | XP_007646701.1 | 13 - 14 | 1075 | 68 | ||
More Information
Set Membership: |
|

Gk
Genomic Map