Assay Details
| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643395.1 | XP_007641585.1 | 3 - 4 | 449 | 65 | |
| XM_007643396.1 | XP_007641586.1 | 3 - 4 | 410 | 65 | ||
| XM_007643397.1 | XP_007641587.1 | 3 - 4 | 424 | 65 | ||
More Information
Set Membership: |
|

Jdp2
Genomic Map