Assay Details
| Gene Symbol: Hmgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644863.1 | XP_007643053.1 | 640 | 82 | ||
| GenBank mRNA | JP048944.1 | - | 640 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmgb2| Gene Symbol: Hmgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644863.1 | XP_007643053.1 | 640 | 82 | ||
| GenBank mRNA | JP048944.1 | - | 640 | 82 | ||
Set Membership: |
|