Assay Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497509.1 | XP_003497557.1 | 4 - 5 | 1124 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fscn3| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497509.1 | XP_003497557.1 | 4 - 5 | 1124 | 68 | |
Set Membership: |
|