Assay Details
| Gene Symbol: Glyctk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497634.2 | XP_003497682.1 | 4 - 5 | 806 | 61 | |
| XM_007615058.1 | XP_007613248.1 | 4 - 5 | 1302 | 61 | ||
More Information
Set Membership: |
|

Glyctk
Genomic Map