Assay Details
| Gene Symbol: LOC100764921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497850.2 | XP_003497898.1 | 3 - 4 | 695 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764921| Gene Symbol: LOC100764921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497850.2 | XP_003497898.1 | 3 - 4 | 695 | 95 | |