Assay Details
| Gene Symbol: LOC100764921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497850.2 | XP_003497898.1 | 4 - 5 | 838 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764921| Gene Symbol: LOC100764921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497850.2 | XP_003497898.1 | 4 - 5 | 838 | 93 | |
Set Membership: |
|