Assay Details
| Gene Symbol: Msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499864.2 | XP_003499912.1 | 5 - 6 | 862 | 97 | |
| XM_007640617.1 | XP_007638807.1 | 4 - 5 | 717 | 97 | ||
| GenBank mRNA | JI879859.1 | - | 3 - 4 | 427 | 97 | |
| JP058986.1 | - | 3 - 4 | 427 | 97 | ||
More Information
Set Membership: |
|

Msmo1
Genomic Map