Assay Details
| Gene Symbol: Hirip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510112.2 | XP_003510160.1 | 5 - 6 | 1822 | 58 | |
| XM_007650741.1 | XP_007648931.1 | 4 - 5 | 1489 | 58 | ||
| GenBank mRNA | JI877625.1 | - | 1 - 2 | 126 | 58 | |
| JP064558.1 | - | 1 - 2 | 126 | 58 | ||
More Information
Set Membership: |
|

Hirip3
Genomic Map