Assay Details
| Gene Symbol: LOC100765753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510465.2 | XP_003510513.1 | 12 - 13 | 1443 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765753| Gene Symbol: LOC100765753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510465.2 | XP_003510513.1 | 12 - 13 | 1443 | 72 | |
Set Membership: |
|