Assay Details
| Gene Symbol: LOC100760251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514130.2 | XP_003514178.2 | 2 - 3 | 96 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760251| Gene Symbol: LOC100760251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514130.2 | XP_003514178.2 | 2 - 3 | 96 | 89 | |
Set Membership: |
|