Assay Details
| Gene Symbol: LOC100763195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642317.1 | XP_007640507.1 | 3 - 4 | 308 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763195| Gene Symbol: LOC100763195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642317.1 | XP_007640507.1 | 3 - 4 | 308 | 96 | |
Set Membership: |
|