Assay Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007629781.1 | XP_007627971.1 | 4 - 5 | 743 | 87 | |
| XM_007630414.1 | XP_007628604.1 | 4 - 5 | 487 | 87 | ||
| GenBank mRNA | JI864912.1 | - | 4 - 5 | 489 | 87 | |
More Information
Set Membership: |
|

Idi1
Genomic Map