Assay Details
| Gene Symbol: Atmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649401.1 | XP_007647591.1 | 2 - 3 | 206 | 84 | |
| GenBank mRNA | JI896008.1 | - | 1 - 2 | 121 | 84 | |
More Information
Set Membership: |
|

Atmin
Genomic Map