Assay Details
| Gene Symbol: LOC100762730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645325.1 | XP_007643515.1 | 4 - 5 | 389 | 89 | |
| XM_007645326.1 | XP_007643516.1 | 4 - 5 | 386 | 89 | ||
More Information
Set Membership: |
|

LOC100762730
Genomic Map