Assay Details
| Gene Symbol: Znf280d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495032.2 | XP_003495080.1 | 10 - 11 | 1249 | 65 | |
| XM_007647595.1 | XP_007645785.1 | 11 - 12 | 1309 | 65 | ||
| XM_007647648.1 | XP_007645838.1 | 11 - 12 | 1362 | 65 | ||
| XM_007647680.1 | XP_007645870.1 | 10 - 11 | 1249 | 65 | ||
| XM_007647720.1 | XP_007645910.1 | 10 - 11 | 1249 | 65 | ||
| GenBank mRNA | JI874670.1 | - | 10 - 11 | 1287 | 65 | |
| JP051087.1 | - | 10 - 11 | 1286 | 65 | ||
More Information
Set Membership: |
|

Znf280d
Genomic Map