Assay Details
| Gene Symbol: Rab18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498903.2 | XP_003498951.1 | 1 - 2 | 261 | 78 | |
| XM_007634997.1 | XP_007633187.1 | 1 - 2 | 262 | 78 | ||
| GenBank mRNA | JI877239.1 | - | 1 - 2 | 136 | 78 | |
| JP054716.1 | - | 1 - 2 | 136 | 78 | ||
More Information
Set Membership: |
|

Rab18
Genomic Map