Assay Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500768.2 | XP_003500816.1 | 3 - 4 | 197 | 62 | |
| XM_007641501.1 | XP_007639691.1 | 3 - 4 | 207 | 62 | ||
| GenBank mRNA | JI866480.1 | - | 2 - 3 | 133 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007641501.1 transcripts.
Set Membership: |
|

Nrbf2
Genomic Map