Assay Details
| Gene Symbol: Cbfa2t2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501119.2 | XP_003501167.1 | 8 - 9 | 1272 | 65 | |
| XM_007641809.1 | XP_007639999.1 | 8 - 9 | 1273 | 65 | ||
More Information
Set Membership: |
|

Cbfa2t2
Genomic Map