Assay Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501867.2 | XP_003501915.2 | 9 - 10 | 1122 | 65 | |
| GenBank mRNA | JI894133.1 | - | 9 - 10 | 1122 | 65 | |
More Information
Set Membership: |
|

Psmc5
Genomic Map