Assay Details
| Gene Symbol: Rpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503886.2 | XP_003503934.1 | 3 - 4 | 512 | 111 | |
| GenBank mRNA | JI862980.1 | - | 3 - 4 | 382 | 111 | |
| JP045283.1 | - | 3 - 4 | 382 | 111 | ||
More Information
Set Membership: |
|

Rpa3
Genomic Map