Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504778.2 | XP_003504826.1 | 4 - 5 | 554 | 100 | |
| XM_007645287.1 | XP_007643477.1 | 4 - 5 | 542 | 100 | ||
| XM_007645288.1 | XP_007643478.1 | 4 - 5 | 555 | 100 | ||
| GenBank mRNA | JI893140.1 | - | 4 - 5 | 553 | 100 | |
| JP047740.1 | - | 4 - 5 | 553 | 100 | ||
More Information
Set Membership: |
|

Sqrdl
Genomic Map