Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504778.2 | XP_003504826.1 | 5 - 6 | 751 | 63 | |
| XM_007645287.1 | XP_007643477.1 | 5 - 6 | 739 | 63 | ||
| XM_007645288.1 | XP_007643478.1 | 5 - 6 | 752 | 63 | ||
More Information
Set Membership: |
|

Sqrdl
Genomic Map