Assay Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504778.2 | XP_003504826.1 | 7 - 8 | 1153 | 132 | |
| XM_007645287.1 | XP_007643477.1 | 7 - 8 | 1141 | 132 | ||
| XM_007645288.1 | XP_007643478.1 | 7 - 8 | 1154 | 133 | ||
| GenBank mRNA | JI893139.1 | - | 2 - 3 | 394 | 132 | |
| JP059776.1 | - | 2 - 3 | 395 | 132 | ||
More Information
Set Membership: |
|

Sqrdl
Genomic Map