Assay Details
| Gene Symbol: Znf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504952.2 | XP_003505000.1 | 5 - 6 | 1103 | 88 | |
| XM_007645447.1 | XP_007643637.1 | 4 - 5 | 1205 | 88 | ||
| XM_007645448.1 | XP_007643638.1 | 3 - 4 | 874 | 88 | ||
| XM_007645449.1 | XP_007643639.1 | 2 - 3 | 822 | 88 | ||
| GenBank mRNA | JP060487.1 | - | 5 - 6 | 1061 | 88 | |
More Information
Set Membership: |
|

Znf131
Genomic Map