Assay Details
Gene Symbol: Efcab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507353.2 | XP_003507401.1 | 2 - 3 | 640 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
