Assay Details
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509221.2 | XP_003509269.1 | 7 - 8 | 1934 | 66 | |
| XM_007649810.1 | XP_007648000.1 | 8 - 9 | 2090 | 66 | ||
| XM_007649811.1 | XP_007648001.1 | 8 - 9 | 2066 | 66 | ||
More Information
Set Membership: |
|

Phc1
Genomic Map