Assay Details
| Gene Symbol: Des | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510595.2 | XP_003510643.1 | 5 - 6 | 1026 | 67 | |
| GenBank mRNA | JI873131.1 | - | 4 - 5 | 560 | 67 | |
More Information
Set Membership: |
|

Des
Genomic Map