Assay Details
| Gene Symbol: Plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515672.2 | XP_003515720.1 | 9 - 10 | 1706 | 71 | |
| GenBank mRNA | JI887396.1 | - | 3 - 4 | 423 | 71 | |
| JP042676.1 | - | 3 - 4 | 424 | 71 | ||
More Information
Set Membership: |
|

Plk1
Genomic Map