Assay Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612568.1 | XP_007610758.1 | 4 - 5 | 783 | 67 | |
| GenBank mRNA | JP047629.1 | - | 4 - 5 | 375 | 67 | |
More Information
Set Membership: |
|

Trim44
Genomic Map