Assay Details
| Gene Symbol: LOC100762617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641838.1 | XP_007640028.1 | 2 - 3 | 1044 | 82 | |
| XM_007641839.1 | XP_007640029.1 | 3 - 4 | 598 | 82 | ||
| XM_007641840.1 | XP_007640030.1 | 2 - 3 | 1044 | 82 | ||
| XM_007641841.1 | XP_007640031.1 | 2 - 3 | 873 | 82 | ||
More Information
Set Membership: |
|

LOC100762617
Genomic Map