Assay Details
| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650650.1 | XP_007648840.1 | 1 - 2 | 205 | 73 | |
| GenBank mRNA | JI867430.1 | - | 1 - 2 | 205 | 73 | |
More Information
Set Membership: |
|

Gtf2b
Genomic Map