Assay Details
| Gene Symbol: Wdr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652651.1 | XP_007650841.1 | 2 - 3 | 2674 | 55 | |
| GenBank mRNA | JP041824.1 | - | 2 - 3 | 1339 | 55 | |
More Information
Set Membership: |
|

Wdr6
Genomic Map