Assay Details
| Gene Symbol: Prkab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495911.2 | XP_003495959.1 | 6 - 7 | 943 | 64 | |
| XM_007645145.1 | XP_007643335.1 | 6 - 7 | 946 | 64 | ||
| XM_007645152.1 | XP_007643342.1 | 7 - 8 | 841 | 64 | ||
| GenBank mRNA | JP063562.1 | - | 2 - 3 | 210 | 64 | |
More Information
Set Membership: |
|

Prkab1
Genomic Map