Assay Details
| Gene Symbol: LOC100768660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511992.2 | XP_003512040.1 | 5 - 6 | 893 | 77 | |
| XM_007652507.1 | XP_007650697.1 | 5 - 6 | 897 | 77 | ||
| GenBank mRNA | JP053044.1 | - | 2 - 3 | 225 | 77 | |
More Information
Set Membership: |
|

LOC100768660
Genomic Map