Assay Details
| Gene Symbol: LOC100761828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515674.2 | XP_003515722.2 | 1 - 2 | 388 | 73 | |
| GenBank mRNA | JI896810.1 | - | 1 - 2 | 215 | 73 | |
| JP057458.1 | - | 1 - 2 | 215 | 73 | ||
More Information
Set Membership: |
|

LOC100761828
Genomic Map