Assay Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642664.1 | XP_007640854.1 | 5 - 6 | 827 | 67 | |
| XM_007642665.1 | XP_007640855.1 | 6 - 7 | 795 | 67 | ||
More Information
Set Membership: |
|

Ripk3
Genomic Map