Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646862.1 | XP_007645052.1 | 4 - 5 | 1202 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmral1| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646862.1 | XP_007645052.1 | 4 - 5 | 1202 | 64 | |
Set Membership: |
|