Assay Details
| Gene Symbol: Cc2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497772.1 | XP_003497820.1 | 13 - 14 | 1568 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cc2d2b| Gene Symbol: Cc2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497772.1 | XP_003497820.1 | 13 - 14 | 1568 | 62 | |
Set Membership: |
|