Assay Details
| Gene Symbol: Ifltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498809.1 | XP_003498857.1 | 7 - 8 | 1222 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifltd1| Gene Symbol: Ifltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498809.1 | XP_003498857.1 | 7 - 8 | 1222 | 58 | |
Set Membership: |
|