Assay Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503604.2 | XP_003503652.1 | 3 - 4 | 563 | 72 | |
| XM_007644197.1 | XP_007642387.1 | 3 - 4 | 563 | 72 | ||
| GenBank mRNA | JI872385.1 | - | 3 - 4 | 518 | 72 | |
More Information
Set Membership: |
|

Nudt21
Genomic Map