Assay Details
| Gene Symbol: LOC100770177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508299.2 | XP_003508347.1 | 3 - 4 | 1454 | 69 | |
| XM_007648928.1 | XP_007647118.1 | 3 - 4 | 1311 | 69 | ||
| XM_007648929.1 | XP_007647119.1 | 3 - 4 | 1620 | 69 | ||
More Information
Set Membership: |
|

LOC100770177
Genomic Map