Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510987.2 | XP_003511035.1 | 1 - 2 | 244 | 62 | |
| GenBank mRNA | JI870153.1 | - | 1 - 2 | 97 | 62 | |
| JP039373.1 | - | 1 - 2 | 97 | 62 | ||
More Information
Set Membership: |
|

Kat8
Genomic Map