Assay Details
Gene Symbol: LOC100754866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007612479.1 | XP_007610669.1 | 2 - 3 | 274 | 80 | |
XM_007612484.1 | XP_007610674.1 | 2 - 3 | 274 | 80 | ||
GenBank mRNA | JP041754.1 | - | 2 - 3 | 284 | 80 |
