Assay Details
| Gene Symbol: LOC100754866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612479.1 | XP_007610669.1 | 3 - 4 | 428 | 77 | |
| XM_007612484.1 | XP_007610674.1 | 3 - 4 | 428 | 77 | ||
| GenBank mRNA | JP041754.1 | - | 3 - 4 | 438 | 77 | |
More Information
Set Membership: |
|

LOC100754866
Genomic Map