Assay Details
| Gene Symbol: Aurkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654371.1 | XP_007652561.1 | 762 | 69 | ||
| GenBank mRNA | JI864587.1 | - | 762 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aurkc| Gene Symbol: Aurkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654371.1 | XP_007652561.1 | 762 | 69 | ||
| GenBank mRNA | JI864587.1 | - | 762 | 69 | ||
Set Membership: |
|