Assay Details
| Gene Symbol: Gid8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503121.2 | XP_003503169.1 | 1 - 2 | 146 | 71 | |
| GenBank mRNA | JI865236.1 | - | 1 - 2 | 134 | 71 | |
| JP044363.1 | - | 1 - 2 | 134 | 71 | ||
More Information
Set Membership: |
|

Gid8
Genomic Map