Assay Details
| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504652.2 | XP_003504700.1 | 7 - 8 | 1068 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hap1| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504652.2 | XP_003504700.1 | 7 - 8 | 1068 | 68 | |
Set Membership: |
|